Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hermansky-Pudlak syndrome 3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89008337 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-008-337 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-008-337 Supplier Abnova Corporation Supplier No. H00084343B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human HPS3 protein.

This gene encodes a protein containing a potential clathrin-binding motif, consensus dileucine signals, and tyrosine-based sorting signals for targeting to vesicles of lysosomal lineage. The encoded protein may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. Mutations in this gene are associated with Hermansky-Pudlak syndrome type 3. Alternate splice variants exist, but their full length sequence has not been determined. [provided by RefSeq

Sequence: MFYVAEPKQVPHILCSPSMKNINPLTAMSYLRKLDTSGFSSILVTLTKAAVALKMGDLDMHRNEMKSHSEMKLVCGFILEPRLLIQQRKGQIVPTELALHLKETQPGLLVASVLGLQKNNKIGIEEADSFFKVLCAKDEDTIPQLLVDFWEAQLVACLPDVVLQELFFKLTSQYIWRLSKRQPPDTTPLRTSEDLINACSHYGLIYPWVHVVISSDSLADKNYTEDLSKLQSLICGPSFDIASIIPFLEPLSEDTIAGLSVHVLCRTRLKEYEQCIDILLERCPEAVIPYANHELKEENRTLWWKKLLPELCQRIKCGGEKYQLYLSSLKETLSIVAVELELKDFMNVLPEDGTATFFLPYLLYCSRKKPLT

Specifications

Antigen Hermansky-Pudlak syndrome 3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human HPS3 protein.
Formulation No additive
Gene HPS3
Gene Accession No. BC016901.1
Gene Alias DKFZp686F0413/FLJ22704/SUTAL
Gene Symbols HPS3
Host Species Mouse
Immunogen HPS3 (AAH16901.1, 1 a.a. ∼ 372 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 84343
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.