Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-HPSE Clone: 4D7,

Catalog No. ST1611100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ST1611100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ST1611100UG Supplier MilliporeSigma™ Supplier No. ST1611100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects HPSE Clone: 4D7 in Human samples, and it is validated for ELISA, Immuno Blotting

Heparanases are endo β-D-glucuronidase enzymes that cleave heparan sulfate side chains of heparan sulfate proteoglycans on cell surfaces and the extracellular matrix. Heparanases are involved in the clotting pathway and interact with tissue factor (TF) and TF pathway inhibitor (TFPI) to promote coagulation. Additional physiological roles include heparan sulfate turnover, embryo development, hair growth, and wound healing. Overexpression of heparanases is strongly implicated in tumor metastasis and angiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HPSE
Applications ELISA, Immunoblot, Immunoblot
Classification Monoclonal
Clone 4D7
Conjugate Unconjugated
Formulation In PBS, pH 7.2.
Host Species Mouse
Immunogen A recombinant polypeptide corresponding to amino acids 72-171 of human HPSE, expressed as a GST fusion protein
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the HPSE protein.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.