Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HRI Rabbit anti-Human, Mouse, Rabbit, Polyclonal, MilliporeSigma™

Catalog No. 07728MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-728-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-728-MI Supplier MilliporeSigma Supplier No. 07728
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect HRI using this Anti-HRI Antibody validated for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HRI
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene Symbols EIF2AK1; KIAA1369; HCR; HRI
Host Species Rabbit
Immunogen 6His-tagged fusion protein corres-ponding to residues 1-138 of mouse heme-regulated inhibitor (HRI)
Purification Method Serum
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_014413.2
Target Species Human, Mouse, Rabbit
Content And Storage −20°C for 24 months
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.