Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hRNF20 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. ABE473MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE473MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE473MI Supplier MilliporeSigma Supplier No. ABE473
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This hRNF20 antibody is validated for use in western blotting for the detection of the hRNF20 protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen hRNF20
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols RNF20; BRE1A
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to human hRNF20.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_062538
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.