Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HS1 Mouse anti-Human, Unlabeled, Clone: 9, BD

Catalog No. BDB610541 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610541 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610541 Supplier BD Biosciences Supplier No. 610541

Mouse Monoclonal Antibody

HS1 (lck-binding protein 1 (lckBP1)) is a protein of 486 amino acids with homology to cytoplasmic tyrosine kinases. The protein contains an SH3 domain and two proline-rich regions, that may be the binding site for lck. Co- mmunoprecipitation experiments demonstrated a tight association between HS1 and lck. The SH3 domain of lck is necessary for this interaction. In addition, HS1 contains tandem repeat sequence of seven amino acids which is a substrate for the tyrosine kinase src. This sequence is also found in the protein cortactin. Therefore, HS1, like cortactin, may be an important adaptor that is critical for the regulation of cellular tyrosine kinases.

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen HS1
Applications Western Blot
Classification Monoclonal
Clone 9
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.