Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

heparan sulfate 2-O-sulfotransferase 1 (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89006826 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-006-826 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-006-826 Supplier Abnova Corporation Supplier No. H00009653B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human HS2ST1 protein.

Heparan sulfate biosynthetic enzymes are key components in generating a myriad of distinct heparan sulfate fine structures that carry out multiple biologic activities. This gene encodes a member of the heparan sulfate biosynthetic enzyme family that transfers sulfate to the 2 position of the iduronic acid residue of heparan sulfate. The disruption of this gene resulted in no kidney formation in knockout embryonic mice, indicating that the absence of this enzyme may interfere with the signaling required for kidney formation. Two alternatively spliced transcript variants that encode different proteins have been found for this gene. [provided by RefSeq

Sequence: MGLLRIMMPPKLQLLAVVAFAVAMLFLENQIQKLEESRSKLERAIARHEVREIEQRHTMDGPRQDATLDEEEDMVIIYNRVPKTASTSFTNIAYDLCAKNKYHVLHINTTKNNPVMSLQDQVRFVKNITSWKEMKPGFYHGHVSYLDFAKFGVKKKPIYINVIRDPIERLVSYYYFLRFGDDYRPGLRRRKQGDKKTFDECVAEGGSDCAPEKLWLQIPFFCGHSSECW

Specifications

Antigen heparan sulfate 2-O-sulfotransferase 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human HS2ST1 protein.
Formulation No additive
Gene HS2ST1
Gene Accession No. BC025990.1
Gene Alias FLJ11317/KIAA0448/MGC131986/dJ604K5.2
Gene Symbols HS2ST1
Host Species Mouse
Immunogen HS2ST1 (AAH25990.1, 1 a.a. ∼ 229 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 9653
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.