Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hydroxysteroid (11-beta) dehydrogenase 1, Mouse, Clone: 2C10, Abnova™

Catalog No. 89001461 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-461 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-461 Supplier Abnova Corporation Supplier No. H00003290M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant HSD11B1.

The protein encoded by this gene is a microsomal enzyme that catalyzes the conversion of the stress hormone cortisol to the inactive metabolite cortisone. In addition, the encoded protein can catalyze the reverse reaction, the conversion of cortisone to cortisol. Too much cortisol can lead to central obesity, and a particular variation in this gene has been associated with obesity and insulin resistance in children. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq

Sequence: MAFMKKYLLPILGLFMAYYYYSANEEFRPEMLQGKKVIVTGASKGIGREMAYHLAKMGAHVVVTARSKETLQKVVSHCLELGAASAHYIAGTMEDMTFAEQFVAQAGKLMGGLDMLILNHITNTSLNLFHDDIHHVRKSMEVNFLSYVVLTVAALPMLKQSNGSIVVVSSLAGKVAYPMVAAYSASKFALDGFFSSIRKEYSVSRVNVSITLCVLGLIDTETAMKAVSGIVHMQAAPKEECALEIIKGGALRQEEVYYDSSLWTTLLIRNPCRKILEFLYSTSYNMDRFINK

Specifications

Antigen hydroxysteroid (11-beta) dehydrogenase 1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2C10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant HSD11B1.
Formulation ascites with no preservative
Gene HSD11B1
Gene Accession No. BC012593
Gene Alias 11-DH/11-beta-HSD1/HDL/HSD11/HSD11B/HSD11L/MGC13539/SDR26C1
Gene Symbols HSD11B1
Host Species Mouse
Immunogen HSD11B1 (AAH12593, 1 a.a. ∼ 292 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 3290
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.