Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hydroxysteroid (17-beta) dehydrogenase 12 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89004621 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-621 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-621 Supplier Abnova Corporation Supplier No. H00051144A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant HSD17B12.

The enzyme 17-beta hydroxysteroid dehydrogenase-12 (HSD17B12) uses NADPH to reduce 3-ketoacyl-CoA to 3-hydroxyacyl-CoA during the second step of fatty acid elongation.[supplied by OMIM

Sequence: SATKTFVDFFSQCLHEEYRSKGVFVQSVLPYFVATKLAKIRKPTLDKPSPETFVKSAIKTVGLQSRTNG

Specifications

Antigen hydroxysteroid (17-beta) dehydrogenase 12
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant HSD17B12.
Formulation 50% glycerol
Gene HSD17B12
Gene Accession No. NM_016142
Gene Alias KAR/SDR12C1
Gene Symbols HSD17B12
Host Species Mouse
Immunogen HSD17B12 (NP_057226, 203 a.a. ∼ 271 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 51144
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.