Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSD17B12 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526360
Description
This antibody is predicted to react with non-human primate based on sequence homology.
The enzyme 17-beta hydroxysteroid dehydrogenase-12 (HSD17B12) uses NADPH to reduce 3-ketoacyl-CoA to 3-hydroxyacyl-CoA during the second step of fatty acid elongation.
Specifications
HSD17B12 | |
Polyclonal | |
HSD17B12 | |
KAR, SDR12C1, 17-beta-HSD 12, 17-beta-hydroxysteroid dehydrogenase 12, 17beta-HSD type 12, 3-ketoacyl-CoA reductase, estradiol 17-beta-dehydrogenase 12, short chain dehydrogenase/reductase family 12C, member 1, steroid dehydrogenase homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51144 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
Q53GQ0 | |
HSD17B12 | |
KLH conjugated synthetic peptide between 126-155 amino acids from the central region of human HSD17B12 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction