Learn More
hydroxysteroid (17-beta) dehydrogenase 3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant HSD17B3.
Supplier: Abnova Corporation H00003293A01
Description
This isoform of 17 beta-hydroxysteroid dehydrogenase is expressed predominantly in the testis and catalyzes the conversion of androstenedione to testosterone. It preferentially uses NADP as cofactor. Deficiency can result in male pseudohermaphroditism with gynecomastia. [provided by RefSeq
Sequence: RCVLLNYWKVLPKSFLRSMGQWAVITGAGDGIGKAYSFELAKRGLNVVLISRTLEKLEAIATEIERTTGRSVKIIQADFTKDDIYEHIKEKSpecifications
hydroxysteroid (17-beta) dehydrogenase 3 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant HSD17B3. | |
HSD17B3 | |
EDH17B3/SDR12C2 | |
Mouse | |
50 μL | |
Yes | |
3293 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_000197 | |
HSD17B3 | |
HSD17B3 (NP_000188, 29 a.a. ∼ 119 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human, Mouse | |
Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.