Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hsp110 Mouse, Unlabeled, Clone: 21, BD

Catalog No. BDB610511 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610511 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610511 Supplier BD Biosciences Supplier No. 610511

Mouse Monoclonal Antibody

With the exception of Hsp110, the stress or heat shock proteins have been thoroughly characterized. Hsp110 has been reported to be expressed in most tissues. Like other stress proteins, Hsp110 is induced by heat shock and its induction is associated with thermotolerance. The sequence of Hsp110 reportedly bears similarity to several Hsp70-related proteins that have been termed Hsp110/SSE. The carboxy-terminal domains of these proteins and Hsp110 contain a putative peptide-binding site. These proteins also contain six highly conserved regions found in the same progression, as well as five conserved ATP-binding motifs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hsp110
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 21
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.