Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP27 Rabbit anti-Hamster, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 06517MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-517-MI 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-517-MI Supplier MilliporeSigma Supplier No. 06517

Rabbit Polyclonal Antibody

Anti-HSP27 Antibody, CT is an antibody against HSP27 for use in western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSP27
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide, before the addition of glycerol to 30%
Gene Symbols HSPB1; Hs.76067; HSP27; Hsp25; HspB1; HSP28; CMT2F; HS.76067; DKFZp586P1322; HMN2B; SRP27
Host Species Rabbit
Immunogen Synthetic peptide (AGKSEQSGAK) corresponding to the C-terminal sequence of hamster HSP27
Purification Method Protein A chromatography
Quantity 250 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001540
Target Species Hamster, Mouse, Rat
Content And Storage −20°C for 24 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.