Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat HSP40/DNAJB1 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$514.50

Specifications

Antigen HSP40/DNAJB1
Clone 419401
Classification Monoclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB4145
GREENER_CHOICE
View Documents
R&D Systems
MAB4145
100 μg
Each for $514.50
Only null left
Add to Cart
 
A410
View Documents
R&D Systems
A410
50 μg N/A N/A N/A
Description

Description

HSP40/DNAJB1 Monoclonal specifically detects HSP40/DNAJB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Knockout Validated.
Specifications

Specifications

HSP40/DNAJB1
Monoclonal
Purified
RUO
DnaJ (Hsp40) homolog, subfamily B, member 1, DnaJ (Hsp40) homolog, subfmaily B, member 1, dnaJ homolog subfamily B member 1, DnaJ protein homolog 1, DNAJ1, DNAJB1, hDj-1, HDJ1, Heat shock 40 kDa protein 1, heat shock 40kD protein 1, Heat shock protein 40, HSP40, HSPF1, HSPF1Hdj1, Human DnaJ protein 1, radial spoke 16 homolog B, RSPH16B, Sis1
DNAJB1
Primary
419401
Unconjugated
Mouse
Human, Mouse, Rat
3337
IgG2b
Protein A or G purified from hybridoma culture supernatant
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.