Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Hsp47 Clone: M16.10A1,

Catalog No. 38602350UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
38-602-350UG 50 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-602-350UG Supplier MilliporeSigma™ Supplier No. 38602350UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Hsp47 Clone: M16.10A1 in Bovine, Canine, Chicken, Guinea Pig, Hamster, Human, Monkey, Mouse, Porcine, Rat, Sheep samples, and it is validated for ELISA, Enzyme Immunoassay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immuno Blotting

Hsp47 is a unique heat shock protein with collagen binding ability. A member of the serpin family of proteins, Hsp47 localizes within the ER due to an RDEL sequence, a variant of the KDEL sequence. Hsp47 is thought to bind procollagen immediately after it co-translationally enters the ER, and it dissociates from procollagen before its secretion. The ability of Hsp47 to bind collagen I and collagen IV suggests it may function as a molecular chaperone for these proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hsp47
Applications ELISA, Immunoblot, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone M16.10A1
Conjugate Unconjugated
Formulation In PBS, 50% glycerol, pH 7.2.
Host Species Mouse
Immunogen Hsp47 from L6 cells
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽47kDa Hsp47 protein.
Target Species Bovine, Canine, Chicken, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rat, Sheep
Content And Storage −20°C
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.