Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hsp70 Mouse, Unlabeled, Clone: 7, BD

Catalog No. BDB610607 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610607 50 μg
BDB610608 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610607 Supplier BD Biosciences Supplier No. 610607
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hsp70 is a member of the heat shock or stress-induced genes whose expression is regulated at both transcriptional and translational levels. Elevated temperatures induce a transient resistance to subsequent heat exposure in various types of cells. Although Hsp70 protein is constitutively expressed at low levels, its expression abundantly increases in response to heat stress. During such stress, Hsp70 leaves the cytoplasm and nucleoplasm and rapidly accumulates in the nucleolus. Removal of the stress results in the redistribution of Hsp70 to its original locations.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Hsp70
Applications Western Blot
Classification Monoclonal
Clone 7
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.