Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Hsp70 Clone: C92F3A-5,

Catalog No. 38603250UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
38-603-250UG 50 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-603-250UG Supplier MilliporeSigma™ Supplier No. 38603250UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Hsp70 Clone: C92F3A-5 in Multi samples, and it is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Immuno Blotting

The Hsp70 family consists of multiple members, all of which bind ATP in vitro, that are localized within different intracellular compartments. Hsp70 is not usually expressed under normal growth conditions, but is expressed at high levels in cells experiencing stress. It is found in the cytosol, nucleus, and nucleolus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hsp70
Applications ELISA, Flow Cytometry, Immunoblot, Immunoblot, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone C92F3A-5
Conjugate Unconjugated
Formulation In PBS, 50% glycerol, pH 7.2.
Host Species Mouse
Immunogen Hsp70 from HeLa cells
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽70kDa Hsp70 protein. Does not cross-react with Hsc70.
Target Species Multi-species
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.