Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP70/HSPA1A Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 6 publications
Supplier: Novus Biologicals NBP177456SS
Description
HSP70/HSPA1A Polyclonal specifically detects HSP70/HSPA1A in Human, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Electron Microscopy, Block/Neutralize.Specifications
| HSP70/HSPA1A | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| dnaK-type molecular chaperone HSP70-1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock 70kDa protein 1A, heat shock-induced protein, HSP70.1/HSP70.2, HSP70-1, HSP70-1/HSP70-2, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1B | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay, Neutralization | |
| 1.0 mg/mL | |
| Western Blot 1:1000, Simple Western 1:50, Flow Cytometry reported in scientific literature (PMID 33760187), Immunohistochemistry 1:400, Immunohistochemistry-Paraffin 1:400, Electron Microscopy reported in scientific literature (PMID 33335563), Block/Neutralize reported in scientific literature (PMID 30417859) | |
| P08107 | |
| HSPA1A | |
| A synthetic peptide made to a C-terminus portion of the human Hsp70 protein (between residues 600-641) [UniProt # P08107] | |
| 0.025 mL | |
| Cancer, Cellular Markers, Hypoxia, Membrane Trafficking and Chaperones | |
| 3303 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction