Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Hsp72/73 Clone: W27,

Catalog No. HSP01100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
HSP01100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. HSP01100UG Supplier MilliporeSigma™ Supplier No. HSP01100UG

Mouse Monoclonal Antibody

Specifically detects Hsp72/73 Clone: W27 in Human, Mouse, Rat samples, and it is validated for Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation

The heat shock response was first described for Drosophila salivary gland cells, and morphologically consists of a change in their polytene chromosome puffing patterns that involves de novo synthesis of a few proteins. Similar heat shock proteins were later discovered in bacterial, chicken, and mammalian cells, and were subsequently studied in most organisms. A series of heatshock proteins Hsp 90, Hsp 70, Hsp 20-30 and ubiquitin (8,000 m.w.) are induced by insults such as temperature shock, chemicals and other environmental stress. Hsp 70 levels increase 1000-fold in mammalian and Drosophila cells following induction. This protein concentrates in nuclei during heat shock and returns to the cytoplasm when the shock is removed. The exact function(s) of the Hsp 70 and other heat shock proteins are not known. Mammalian Hsp 72/73 is reported to be structurally and functionally related to a protein that releases clathrin triskelia from coated vesicles; Hsp 72/73 is known to bind ATP without hydrolyzing it.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hsp72/73
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone W27
Conjugate Unconjugated
Formulation In 0.05M sodium phosphate buffer, 0.2% gelatin.
Host Species Mouse
Immunogen purified Hsp72/73
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽72kDa constitutive and inducible forms of Hsp72/73 in heat-shocked HeLa cells.
Target Species Human, Mouse, Rat
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.