Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hsp75/TRAP1 Mouse, Unlabeled, Clone: 42, BD

Catalog No. BDB612344 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612344 50 μg
BDB612345 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB612344 Supplier BD Biosciences Supplier No. 612344
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The hsp90 family of molecular chaperones is involved in a number of signal transduction pathways, including cell division, protein folding and transport. A new member of this family was recently identified as TRAP1 (Tumor necrosis factor receptor-associated protein) or hsp75. TRAP1 is a mitochondrial protein that is an ATPase and ATP-binding protein, rendering it similar to hsp90. There is a high degree of homology between TRAP1 and hsp90 in 6 domains, however one major difference is that a highly charged domain present in hsp90 is absent in TRAP1. TRAP1 has been shown to interact with the intracellular domain of the type 1 receptor for tumor necrosis factor (TNFR-1IC) and to the retinoblastoma protein (Rb). Signaling through TNFR-1 induces antiviral responses, fibroblast proliferation, cytotoxicity, etc. TRAP1 binds TNFR-1IC outside of the death domain leading to disruption in the signaling by TNFR-1. Rb has been shown to play a pivotal role in cellular differentiation and progression through the cell cycle. TRAP1 associates with Rb via an LxCxE motif, which is common among proteins that interact with Rb through SV40 T-antigen binding domain. Thus, TRAP1 is a new member of the hsp90 family that interacts with Rb and TNFR-1IC.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Hsp75/TRAP1
Applications Western Blot
Classification Monoclonal
Clone 42
Concentration 250μg/mL
Conjugate Unconjugated
Description TRAP1
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Hsp75/TRAP1 aa. 253-464
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.