Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP90 Mouse anti-Bovine, Chicken, Human, Mouse, Pig, Rat, Rabbit, Clone: D7a, MilliporeSigma™

Catalog No. 05594MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-594-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-594-MI Supplier MilliporeSigma Supplier No. 05594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-HSP90 Antibody is validated for use in immunoprecipitation, western blotting for the detection of HSP90.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSP90
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Protein G Purified immunoglobulin in 0.1M Tris-Glycine, 0.15M NaCl, 0.05% Sodium Azide, pH 7.4.
Gene Accession No. NM_005348
Gene Symbols HSP90AA1, HSP90A, Hsp90, HSPCAL1, HSPCAL4, HSPN, HSP86, HSPCA, Hsp89, HSP90N, LAP2, HSPC1, FLJ31884
Host Species Mouse
Immunogen Full-length protein purified from chicken brain; clone D7a
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Chicken, Human, Mouse, Pig, Rabbit, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.