Learn More
HSP90AB1, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human HSP90AB1 peptide using ARM Technology.
Supplier: Abnova Corporation H00003326K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. There are 2 major cytosolic HSP90 proteins, HSP90AA1 (MIM 140571), an inducible form, and HSP90AB1, a constitutive form. Other HSP90 proteins are found in endoplasmic reticulum (HSP90B1; MIM 191175) and mitochondria (TRAP1; MIM 606219) (Chen et al., 2005 [PubMed 16269234]).[supplied by OMIM
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
HSP90AB1 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
HSP90AB1 | |
HSP90AB1 | |
A synthetic peptide of human HSP90AB1 | |
RUO | |
3326 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
heat shock protein 90kDa alpha (cytosolic), class B member 1 | |
HSP90AB1 | |
D6S182, FLJ26984, HSP90-BETA, HSP90B, HSPC2, HSPCB | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.