Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP90 beta Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA515031
Description
This antibody is predicted to react with bovine, chicken, mouse, non-human primate and rat based on sequence homology.
Heat shock proteins (HSP) are expressed in response to various biological stresses, including heat. HSP90 is a 90 kDa protein that is induced under stress conditions, but is also one of the most abundant cellular proteins found under non-stress conditions. HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. HSP90 has been found to be associated with a number of other intracellular proteins, including steroid receptors, actin, tubulin, aryl hydrocarbon (Ah) receptor, and some kinases.
Specifications
HSP90 beta | |
Polyclonal | |
HSP90AB1 | |
D6S182; HSP90-BETA; HSP90B; HSPC2; HSPCB; 90-kda heat shock protein beta HSP90 beta; HSP 84; HSP 90; HSP84; heat shock 84 kDa; heat shock 90kD protein 1; beta; heat shock 90kDa protein 1; beta; heat shock protein HSP 90-beta; heat shock protein beta | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
3326 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
P08238 | |
HSP90AB1 | |
KLH conjugated synthetic peptide between 438-465 amino acids from the central region of human HSP90AB1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction