Learn More
HSP70, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA514521
Description
HSPA1A is a member of the heat shock protein 70 family. In conjunction with other heat shock proteins, this protein stabilizes existing proteins against aggregation and mediates the folding of newly translated proteins in the cytosol and in organelles. It is also involved in the ubiquitin-proteasome pathway through interaction with the AU-rich element RNA-binding protein 1.
This antibody is predicted to react with bovine, chicken, canine, equine, hamster, mouse, porcine, non-human primate, rat and Xenopus, yeast and zebrafish based on sequence homology.
Specifications
HSP70 | |
Polyclonal | |
HSPA1A | |
HSP70-1, HSP70-1A, HSP70I, HSP72, HSPA1, HSP70-1/HSP70-2, HSP70.1/HSP70.2, dnaK-type molecular chaperone HSP70-1, heat shock 70kDa protein 1/2, heat shock 70kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock-induced protein, Hspa70 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3303 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
P0DMV8 | |
HSPA1A | |
KLH conjugated synthetic peptide between 19-48 amino acids from human HSPA1A/HSPA1B | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.