Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSPA5 Mouse anti-Human, Rat, Mouse, Clone: 4E3, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABC675
Description
Anti-HSPA5 Antibody, clone 4E3 is a highly specific mouse monoclonal antibody, that targets GRP-78 and has been tested in western blotting and immunohistochemistry.
Specifications
HSPA5 | |
Monoclonal | |
Unconjugated | |
NP_005338 | |
Mouse | |
Unpurified | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
Immunohistochemistry, Western Blot | |
4E3 | |
Mouse monoclonal IgG2b ascitic fluid containing up to 0.1% sodium azide. | |
HSPA5, GRP78 | |
Purified recominant fragment of human HSPA5 expressed in E. coli. | |
100 μL | |
Human, Mouse, Rat | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction