Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti- HSPA8/Hsc70 Clone: N69,

Catalog No. MABE1120MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABE1120MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1120MI Supplier MilliporeSigma™ Supplier No. MABE1120
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects HSPA8/Hsc70 Clone: N69 in Bovine, Human, Mouse, Rat samples, and it is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blotting

Heat shock cognate 71kDa protein (UniProt P19120; also known as Heat shock 70kDa protein 8, HSC70, HSC71, HSC73) is encoded by the HSPA8 (also known as HSC70, HSC73) gene (Gene ID 281831) in bovine species. The mammalian HSP70 family consists of the mitochondrial HSP75, the endoplasmic HSP78, the cytoplasmic HSC70 (a.k.a. HSC73 and HSP73) and its inducible counterpart HSP72 (a.k.a. HSP70). The latter two have similar structures and properties, and both of them bind misfolded, newly synthesized polypeptides in an ATP-dependent manner, reflecting their chaperonic activity. All cells constitutively express HSC70. On the other hand, the expression of HSP72 in normal cells is mainly stress inducible, although constitutively high HSP72 expressions are observed in malignant human tumors of various origins. The induced HSP72 expression enables stressed cells to better cope with the elevation of unfolded/denatured proteins and prevent apoptosis under various stress conditions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSPA8/Hsc70
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone N69
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P19120
Gene Symbols HSPA8, HSC70
Host Species Mouse
Immunogen Co-purified Hsc73 and Hsp72 proteins from bovine skeletal muscle.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics, Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_776770
Test Specificity Clone N69 reacts with the constitutively expressed HSPA8/Hsc70 (HSC73), but not the heat schock-inducible HSP72 (Demidov, O.N., et al. (1999). Eur. J. Cardiothorac. Surg. 16(4):444-449).
Target Species Bovine, Human, Mouse, Rat
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.