Learn More
Description
The protein encoded by this gene belongs to the superfamily of small heat-shock proteins containing a conservative alpha-crystallin domain at the C-terminal part of the molecule. The expression of this gene in induced by estrogen in estrogen receptor-positive breast cancer cells, and this protein also functions as a chaperone in association with Bag3, a stimulator of macroautophagy. Thus, this gene appears to be involved in regulation of cell proliferation, apoptosis, and carcinogenesis, and mutations in this gene have been associated with different neuromuscular diseases, including Charcot-Marie-Tooth disease. [provided by RefSeq
Specifications
Specifications
| Antigen | HSPB8 |
| Applications | ELISA, Immunohistochemistry (PFA fixed), Western Blot |
| Classification | Monoclonal |
| Clone | 5B12 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant HSPB8. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | HSPB8 |
| Gene Accession No. | NM_014365 |
| Gene Alias | CMT2L/DHMN2/E2IG1/H11/HMN2/HMN2A/HSP22 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.