Learn More
HSPA (heat shock 70kDa) binding protein, cytoplasmic cochaperone 1, Mouse, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
Specifications
Specifications
| Antigen | HSPA (heat shock 70kDa) binding protein, cytoplasmic cochaperone 1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a full-length human HSPBP1 protein. |
| Formulation | No additive |
| Gene | HSPBP1 |
| Gene Accession No. | NM_012267.3 |
| Gene Alias | FES1 |
| Gene Symbols | HSPBP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.