Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSV Tag Antibody - BSA Free, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB600513
Description
HSV Tag Polyclonal antibody specifically detects HSV Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation.Specifications
HSV Tag | |
Polyclonal | |
Unconjugated | |
PBS with 0.09% Sodium Azide | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
IgG |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:1000 - 1:30000, ELISA 1:1000-1:30000 (Primary) 1:100-1:500 (For Coating Plates), Immunocytochemistry/Immunofluorescence 1:100-1:400, Immunoprecipitation 1 - 4 ug/mg lysate | |
HSV epitope tag | |
Goats were immunized with HSV (SQPELAPEDPED) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support. | |
0.1 mL | |
Epitope Tags | |
Store at 4C. Do not freeze. | |
Epitope Tag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction