Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-HTLV-I gp46, Clone: 68/4.11.21, ZeptoMetrix

Murine monoclonal antibody reacts with Human T-Lymphotropic Virus Type I (HTLV-I) surface protein, gp46
Supplier: ZeptoMetrix 0801118
Description
Host Species: Mouse
Clone: 68/4.11.21
Isotype: IgG1
Species Reactivity: Hu
Enzyme Immunoassay, Immunofluorescence, Western Blotting
Specifications
HTLV-I gp46 | |
68/4.11.21 | |
Mouse | |
RUO | |
Human | |
IgG1 |
Monoclonal | |
Unconjugated | |
1 mg | |
Primary | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction