Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HTLV-II p19, Clone: 78/6.18.07, ZeptoMetrix
SDP

Murine monoclonal antibody reacts with Human T-Lymphotropic Virus Type II (HTLV-II) core p19

$376.00 - $2495.00

Specifications

Antigen HTLV-II p19
Clone 78/6.18.07
Classification Monoclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
22-157-339
SDP
View Documents
ZeptoMetrix
0801093
100 μg
Each for $376.00
Only null left
Add to Cart
 
22-157-346
SDP
View Documents
ZeptoMetrix
0801113
1 mg
Each for $2,495.00
Only null left
Add to Cart
 
Description

Description

Host Species: Mouse
Clone: 78/6.18.07
Isotype: IgG2b
Species Reactivity: Hu

Enzyme Immunoassay, Immunofluorescence, Western Blotting

Specifications

Specifications

HTLV-II p19
Monoclonal
Purified
RUO
IgG2b
78/6.18.07
Unconjugated
Mouse
Human
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.