Learn More
HTR3B, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human HTR3B protein.
Supplier: Abnova Corporation H00009177B01P
Description
The product of this gene belongs to the ligand-gated ion channel receptor superfamily. This gene encodes subunit B of the type 3 receptor for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor causes fast, depolarizing responses in neurons after activation. It appears that the heteromeric combination of A and B subunits is necessary to provide the full functional features of this receptor, since either subunit alone results in receptors with very low conductance and response amplitude. [provided by RefSeq
Sequence: MLSSVMAPLWACILVAAGILATDTHHPQDSALYHLSKQLLQKYHKEVRPVYNWTKATTVYLDLFVHAILDVDAENQILKTSVWYQEVWNDEFLSWNSSMFDEIREISLPLSAIWAPDIIINEFVDIERYPDLPYVYVNSSGTIENYKPIQVVSACSLETYAFPFDVQNCSLTFKSILHTVEDVDLAFLRSPEDIQHDKKAFLNDSEWELLSVSSTYSILQSSAGGFAQIQFNVVMRRHPLVYVVSLLIPSIFLMLVDLGSFYLPPNCRARIVFKTSVLVGYTVFRVNMSNQVPRSVGSTPLIGHFFTICMAFLVLSLAKSIVLVKFLHDEQRGGQEQPFLCLRGDTDADRPRVEPRAQRAVVTESSLYGEHLAQPGTLKEVWSQLQSISNYLQTQDQTDQQEAEWLVLLSRFDRLLFQSYLFMLGIYTITLCSLWALWGGVSpecifications
HTR3B | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human HTR3B protein. | |
HTR3B | |
5-HT3B | |
Mouse | |
Affinity chromatography | |
RUO | |
9177 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_006028.3 | |
HTR3B | |
HTR3B (NP_006019.1, 1 a.a. ∼ 441 a.a) full-length human protein. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.