Learn More
HTR5A, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human HTR5A peptide using ARM Technology.
Supplier: Abnova Corporation H00003361K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The neurotransmitter serotonin (5-hydroxytryptamine, 5-HT) has been implicated in a wide range of psychiatric conditions and also has vasoconstrictive and vasodilatory effects. The gene described in this record is a member of 5-hydroxytryptamine (serotonin) receptor family and encodes a multi-pass membrane protein that functions as a receptor for 5-hydroxytryptamine and couples to G-proteins. This protein has been shown to function in part through the regulation of intracellular Ca2+ mobilization. [provided by RefSeq]
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
HTR5A | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
HTR5A | |
HTR5A | |
A synthetic peptide of human HTR5A | |
RUO | |
3361 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
5-hydroxytryptamine (serotonin) receptor 5A | |
HTR5A | |
5-HT5A, MGC138226 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.