Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HU-B27 Mouse anti-Human, Mouse, Rat, FITC, MilliporeSigma™

Catalog No. MAB4212FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4212FMI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4212FMI Supplier MilliporeSigma Supplier No. MAB4212F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect HU-B27d using this Anti-HU-B27 Antibody, FITC conjugated validated for use in flow cytometry, immunocytochemistry, immunofluorescence and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HU-B27
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Conjugate FITC
Formulation The reagent is supplied in 1.0 mL phosphate-buffered saline, pH 7.4, containing 0.2% bovine serum albumin and 0.1% sodium azide. Purified from mouse ascitic fluid. The characteristics of each lot are tested by gel electrophoresis and flow cytometry.
Gene Symbols MRAP, C21orf61, FGD2, FALP, B27, GCCD2
Host Species Mouse
Quantity 1 mL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots in dark for up to 6 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.