Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HuC/HuD Mouse anti-Human, Rat, Mouse, Clone: 15A7.1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABN153
Description
This HuC/HuD antibody is validated for use in western blotting and immunohistochemistry for the detection of the HuC/HuD protein.
Specifications
HuC/HuD | |
Monoclonal | |
Unconjugated | |
NP_001411 | |
Mouse | |
Protein G purified | |
Primary | |
2-8°C for one year | |
IgG2a κ |
Immunohistochemistry, Western Blot | |
15A7.1 | |
Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
ELAVL3, HUC, PLE21, ELAVL4, HUD, PNEM | |
KLH-conjugated linear peptide corresponding to human HuC/HuD. | |
100 μg | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction