Learn More
Description
- Antigen: Human Albumin
- Polyclonal
- Primary Antibody
- Host: Rabbit
- Form: lyophilized (IgG)
- Conjugate: Unlabeled
- Immunoblotting, Immunoassay
- Gives sharp precipitin lines in immunoelectrophoresis and immunodiffusion techniques
- Suitable for conjugation with enzymes, radiolabels, fluorochromes or other markers, and for attachment to solid supports for use as an immunoadsorbant
- Can be used as a primary reagent in enzyme immunoassays, fluorescein immunoassays and blot, cell or tissue immunostaining
Specifications
Specifications
| Antigen | Human Albumin |
| Classification | Polyclonal |
| Concentration | N/A |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Quantity | 2 mL |
| Primary or Secondary | Primary |
| Form | Lyophilized |
| Isotype | IgG |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.