Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin-like Protein 2/ANGPTL2 Goat anti-Human, Polyclonal, R&D Systems™

Catalog No. AF2084
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2084 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2084 Supplier R&D Systems Supplier No. AF2084
This item is not returnable. View return policy

Goat Polyclonal Antibody

Angiopoietin-like Protein 2/ANGPTL2 Polyclonal specifically detects Angiopoietin-like Protein 2/ANGPTL2 in Human samples. It is validated for Western Blot.

Specifications

Antigen Angiopoietin-like Protein 2/ANGPTL2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL
Gene Accession No. Q9UKU9
Gene Alias angiopoietin-like 2, Angiopoietin-like protein 2, angiopoietin-related protein 2, ANGPTL2, ANGRP2, ARP2HARP, MGC8889
Gene Symbols ANGPTL2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-like 2 Gln23-Phe492 Accession # Q9UKU9
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23452
Test Specificity Detects human Angiopoietin-like 2 (ANGPTL2) in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse ANGPTL2 is observed and less than 1% cross-reactivity with recombinant human (rh) ANGPTL6 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.