Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calreticulin Goat anti-Human, Polyclonal, R&D Systems™

Catalog No. AF3898
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3898 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3898 Supplier R&D Systems Supplier No. AF3898
This item is not returnable. View return policy

Goat Polyclonal Antibody

Calreticulin Polyclonal antibody AF3898 Specifically detects Calreticulin in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry, Simple Western, Western Blotting

Specifications

Antigen Calreticulin
Applications Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL
Gene Accession No. P27797
Gene Alias autoantigen Ro, CALR, Calregulin, calreticulin, cC1qR, CRP55, CRT, CRTC, Endoplasmic reticulum resident protein 60, ERp60, FLJ26680, grp60, RO, Sicca syndrome antigen A (autoantigen Ro; calreticulin), SSA, SSAHACBP, Vasostatin
Gene Symbols CALR
Host Species Goat
Immunogen E. coli-derived recombinant human Calreticulin, Met1-Asn180, Accession # P27797
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 811
Test Specificity Detects human Calreticulin in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.