Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-13 Sheep anti-Human, Canine, Polyclonal, R&D Systems™

Catalog No. AF5894
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5894 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5894 Supplier R&D Systems Supplier No. AF5894
This item is not returnable. View return policy

Sheep Polyclonal Antibody

IL-13 Polyclonal antibody AF5894 Specifically detects IL-13 in Canine, Human samples. It is validated for Western Blotting

Specifications

Antigen IL-13
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q9N0W9
Gene Alias ALRHMGC116789, BHR1interleukin-13, IL13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600
Gene Symbols IL13
Host Species Sheep
Immunogen E. coli-derived recombinant canine IL-13, Ser19-Arg131, Accession # Q9N0W9
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3596
Test Specificity Detects recombinant canine IL-13 in direct ELISAs and detects recombinant canine IL-13 and human IL-13 in Western blots. In direct ELISAs, approximately 7% cross-reactivity with recombinant human IL-13 and recombinant mouse IL-13 is observed and less than 2% cross-reactivity with recombinant rat IL-13 and recombinant rhesus monkey IL-13 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human, Canine
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.