Learn More
Description
Host Species: Mouse
Clone: ICRF44
Isotype: IgG1, κ
Cross Reactivity Species: Hu
Immunogen: Human monocytes
Applications:
Flow Cytometry
Specifications
Specifications
| Antigen | Human CD11b/MAC-1 |
| Classification | Monoclonal |
| Clone | ICRF44 |
| Conjugate | Brilliant Violet 605 |
| Host Species | Mouse |
| Immunogen | Human monocytes |
| Purification Method | Affinity Purified |
| Quantity | 100 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.