Learn More
CD127 Mouse anti-Human, Brilliant Violet 605, Clone: HIL-7R-M21, BD
Shop All BD Biosciences ProductsDescription
Host Species: Mouse
Clone: HIL-7R-M21
Isotype: IgG1, κ
Cross Reactivity Species: Hu
Flow Cytometry
Specifications
Specifications
| Antigen | CD127 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | HIL-7R-M21 |
| Conjugate | Brilliant Violet 605 |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 50 Tests |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.