Learn More
Description
Host Species: Mouse
Clone: BNI3
Isotype: IgG2a, κ
Cross Reactivity Species: Hu
Immunogen: Human CTLA4 Recombinant Protein
Applications:
Flow Cytometry
Specifications
Specifications
| Antigen | Human CD152 |
| Classification | Monoclonal |
| Clone | BNI3 |
| Conjugate | PE-CF594 |
| Host Species | Mouse |
| Immunogen | Human CTLA4 Recombinant Protein |
| Purification Method | Affinity Purified |
| Quantity | 50 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.