Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Human CD162, Alexa Fluor 647, Clone: KPL-1; BD
Supplier: BD Biosciences 562758
Description
Host Species: Mouse
Clone: KPL-1
Isotype: IgG1κ
Cross Reactivity Species: Hu
Immunogen: Human PSGL-1 Transfected Cell Line
Flow Cytometry
Specifications
| Human KPL-1 | |
| KPL-1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human |
| Monoclonal | |
| Alexa Fluor 647 | |
| Human PSGL-1 Transfected Cell Line | |
| 50 Tests | |
| Primary |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction