Learn More
Description
Host Species: Mouse
Clone: 5D3
Isotype: IgG2b, κ
Cross Reactivity Species: Hu
Immunogen: Human ABCG2-transfected Mouse cells
Applications:
Flow Cytometry
Specifications
Specifications
| Antigen | Human CD338 |
| Classification | Monoclonal |
| Clone | 5D3 |
| Conjugate | Brilliant Violet 421 |
| Host Species | Mouse |
| Immunogen | Human ABCG2-transfected Mouse cells |
| Purification Method | Affinity Purified |
| Quantity | 50 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.