Learn More
Description
Host Species: Mouse
Clone: B159
Isotype: IgG1κ
Cross Reactivity Species: Hu
Immunogen: Human NK Cells
Flow Cytometry
Specifications
Specifications
| Antigen | Human CD56 |
| Classification | Monoclonal |
| Clone | B159 |
| Conjugate | FITC |
| Host Species | Mouse |
| Immunogen | Human NK Cells |
| Purification Method | Affinity Purified |
| Quantity | 100 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.