Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Human CD56, Clone: NCAM16.2; BV421 Conjugate BD

Catalog No. BDB562752 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562752 25 Tests
BDB562751 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB562752 Supplier BD Biosciences Supplier No. 562752
Add to Cart
Add to Cart

Host Species: Mouse
Clone: NCAM16.2
Isotype: IgG2b, κ
Cross Reactivity Species: Hu
Immunogen: Immunoaffinity-enriched adult human brain NCAM

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Human CD56
Classification Monoclonal
Clone NCAM16.2
Conjugate Brilliant Violet 421
Host Species Mouse
Immunogen Immunoaffinity-enriched adult human brain NCAM
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.