Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chondroitin Sulfate Mouse anti-Human, PE, Clone: 9.2.27, BD

Catalog No. BDB562415 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562415 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562415 Supplier BD Biosciences Supplier No. 562415
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 9.2.27 monoclonal antibody specifically binds to CSPG4 (chondroitin sulfate proteoglycan 4). CSPG4 is also known as MCSP (melanoma chondroitin sulfate proteoglycan) and NG2 (neural/glial antigen 2). The hybridoma secreting the 9.2.27 antibody was generated using a human melanoma cell extract as the immunogen. Tumor cells display a variety of antigens which have been defined by antibodies. Antibodies that specifically react with tumor-associated antigens are useful in understanding the biology of particular tumor types. Proteoglycans including chondroitin sulfate, keratan sulfate, dermatan sulfate and heparan sulfate are major components of the extracellular matrices of many animal cell types. Clone 9.2.27 has been shown to react with human melanoma cells, glioma cells, and proliferating brain endothelial cells. It did not react with fetal melanocytes, neuroblastoma LA-N-1 cells or a variety of carcinoma, lymphoid, and fibroblastoid cell lines. Additionally, neither white nor gray matter from the medulla oblongata, cerebellum or spinal cord of a normal human adult reacted with 9.2.27. Clone 9.2.27 has also been used in functional studies to investigate the role of chondroitin sulfate proteoglycans in human tumor cell systems. It has been shown to block melanoma cell spreading, exhibit antiproliferative effects on glioma cells in vitro and suppress glioma tumor growth in athymic nude mice models.

Host Species: Mouse
Clone: 9.2.27
Isotype: IgG2a
Species Reactivity: Hu

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Chondroitin Sulfate
Applications Flow Cytometry
Classification Monoclonal
Clone 9.2.27
Conjugate PE
Description CSPG4; NG2; Chondroitin sulfate proteoglycan 4; MCSP; MCSPG; MELCSPG; MSK16
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Extracts of Human M21 Melanoma Cells
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.