Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hrk Rabbit anti-Human, Polyclonal, R&D Systems™

Catalog No. AF851
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF851 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF851 Supplier R&D Systems Supplier No. AF851

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Hrk Polyclonal antibody AF851 Specifically detects Hrk in Human samples. It is validated for Western Blotting

Specifications

Antigen Hrk
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Alias activator of apoptosis harakiri, activator of apoptosis Hrk, BCL2-interacting protein, BH3-interacting domain-containing protein 3, BID3, death protein 5, DP5, HARAKIRI, harakiri, BCL2 interacting protein (contains only BH3 domain), harakiri, BCL2-interacting protein (contains only BH3 domain), Neuronal death protein DP5
Gene Symbols HRK
Host Species Rabbit
Immunogen KLH-coupled human Hrk synthetic peptide, RTMWRRRARSR RAPAPGAALPC
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8739
Test Specificity Detects human Hrk in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.