Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Human IFN-alpha A, alpha B2 and alpha F, Polyclonal, R&D Systems™

Catalog No. 311301
Change view
Click to view available options
Quantity:
20,000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
311301 20,000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 311301 Supplier R&D Systems Supplier No. 311301
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Host Species: Rabbit
Species Reactivity:Human

Applications:

Neutralization

Specifications

Antigen IFN-alpha
Applications Neutralization
Classification Polyclonal
Conjugate Unconjugated
Dilution Neutralization
Gene Alias IFNA, IFNA2, IFNA2b, IFNalpha, IFN-alphaA, INFA2, interferon alpha 2
Gene Symbols IFNA2
Host Species Rabbit
Immunogen Recombinant human IFN-alpha
Purification Method Antigen Affinity-purified
Quantity 20,000 U
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3440
Test Specificity Human IFN-alpha
Target Species Human
Content And Storage After receipt, the product may be stored at -20degreesC for short-term use (< 6 months). For long-term storage, we recommend storing the product at -70degreesC or below for retention of full activity. Thaw product vial by incubation in cold tap water until just thawed – the contents of the tube should be apportioned in separate tubes so that freezing and thawing is kept to a minimum. Refreezing should be done on dry ice or in a dry ice/alcohol bath. Further dilution of the product should be in buffers containing protein such as 0.1% bovine serum albumin (BSA).
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.