Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-10R alpha Goat anti-Human, R-PE, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Supplier: R&D Systems FAB6280P
This item is not returnable.
View return policy
Description
IL-10R alpha Polyclonal specifically detects IL-10R alpha in Human samples. It is validated for Flow Cytometry.Specifications
IL-10R alpha | |
Polyclonal | |
Flow Cytometry 10 uL/10^6 cells | |
CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL10R alpha, IL-10R subunit 1, IL-10R1, IL10RA, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha | |
Goat | |
Affinity Purified | |
RUO | |
3587 | |
Human | |
IgG |
Flow Cytometry | |
R-PE | |
Q13651 | |
IL10RA | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-10 R alpha His22-Asn235 Accession # Q13651 | |
100 Tests | |
Primary | |
Detects human IL-10 R alpha in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 10% cross-reactivity with recombinant mouse IL-10 R alpha and less than 5% cross-reactivity with recombinant human (rh) CNTF sR alpha and rhIL-2 sR gamma is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction