Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-6R alpha Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AB227NA
This item is not returnable.
View return policy
Description
IL-6R alpha Polyclonal specifically detects IL-6R alpha in Human samples. It is validated for Western Blot, Neutralization.Specifications
| IL-6R alpha | |
| Polyclonal | |
| Unconjugated | |
| P08887 | |
| IL6R | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-6 R alpha Leu20-Asp358 Accession # P08887 | |
| 1 mg | |
| Primary | |
| Detects human IL-6 R alpha in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant mouse IL-6 R alpha and recombinant human LIF is observed. | |
| Human | |
| Purified |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 1 ug/mL, Neutralization 1-4 ug/mL | |
| CD126, gp80, IL-6 R alpha, IL6Q, IL6R, IL6R alpha, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor | |
| Goat | |
| Protein A or G purified | |
| RUO | |
| 3570 | |
| Reconstitute at 1 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction